CAS RN 60525-15-7
IUPAC Navn:
dihydrogen [(2Z)-3-(4-bromophenyl)-3-(pyridin-3-yl)prop-2-en-1-yl]dimethylamine dichloride
Synonymer:
dihydrogen zimelidine dichloride
Molekylvægt (g/mol):
390.15
Molekylformel:
C16H19BrCl2N2
InChi Key:
CXGURXWCQYHDIR-ULPVBNQHSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CN(C)C\C=C(\C1=CC=C(Br)C=C1)C1=CC=CN=C1
1
–
2
af
2
Resultater
1
Zimelidine Dihydrochloride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Zimelidine Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.