Få mere at vide
Zimelidine Dihydrochloride, Mikromol™
Beskrivelse
Zimelidine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Zimelidine Dihydrochloride |
| CAS | 60525-15-7 |
| Analyt- eller komponentnavne | Zimelidine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H17 Br N2 . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2-Propen-1-amine, 3-(4-bromophenyl)-N,N-dimethyl-3-(3-pyridinyl)-, hydrochloride (1:2), (2Z)-, 2-Propen-1-amine, 3-(4-bromophenyl)-N,N-dimethyl-3-(3-pyridinyl)-, dihydrochloride, (2Z)- (9CI), 2-Propen-1-amine, 3-(4-bromophenyl)-N,N-dimethyl-3-(3-pyridinyl)-, dihydrochloride, (Z)-, H 102/09 hydrochloride, Zimelidine dihydrochloride, Zimelidine hydrochloride |
| InChI formel | InChI=1S/C16H17BrN2.2ClH/c1-19(2)11-9-16(14-4-3-10-18-12-14)13-5-7-15(17)8-6-13;;/h3-10,12H,11H2,1-2H3;2*1H/b16-9-;; |
| SMIL | Cl.Cl.CN(C)C\C=C(\c1ccc(Br)cc1)/c2cccnc2 |
| IUPAC navn | (Z)-3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-3-ylprop-2-en-1-amine;dihydrochloride |
| Vis mere |