CAS RN 603-47-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-{[4-(Dimethylamino)phenyl][4-(methylimino)cyclohexa-2,5-dien-1-ylidene]methyl}-N,N-dimethylaniline Hydrochloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-{[4-(Dimethylamino)phenyl][4-(methylimino)cyclohexa-2,5-dien-1-ylidene]methyl}-N,N-dimethylaniline Hydrochloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-{[4-(Dimethylamino)phenyl][4-(methylimino)cyclohexa-2,5-dien-1-ylidene]methyl}-N,N-dimethylaniline Hydrochloride (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.