CAS RN 60238-56-4
IUPAC Navn:
O-3,4-dichloro-5-(methylsulfanyl)phenyl O,O-diethyl phosphorothioate
Synonymer:
chlorthiophos
Molekylvægt (g/mol):
361.23
Molekylformel:
C11H15Cl2O3PS2
InChi Key:
SBLPWPLKGFWLKK-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)OC1=CC(Cl)=C(Cl)C(SC)=C1
1
–
2
af
2
Resultater
1
Chlorthiophos (Mixture of isomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.