Få mere at vide
Chlorthiophos, SPEX CertiPrep™
Artikelnummer. 16390468 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
16390468 1 mL
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 16390468 Leverandør SPEX Certiprep Leverandørnr. S967
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Chlorthiophos60238-56-41000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Chlorthiophos
Emballage 2mL amber prescored glass ampule
CAS 60238-56-4, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C11H15Cl2O3PS2
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle JAZJVWLGNLCNDD-UHFFFAOYSA-N
SMIL CCOP(=S)(OCC)OC1=CC(=C(C=C1Cl)SC)Cl
IUPAC navn (2,5-dichloro-4-methylsulfanylphenoxy)-diethoxy-sulfanylidene-$l^{5}-phosphane
Opløsningsmiddel eller Matrix Methanol-P&T
Molekylvægt (g/mol) 361.232
PubChem CID 30859
Mængde 1 mL
ChEBI CHEBI:82109
Vis mere Vis mindre
Product intended for laboratory use only