CAS RN 602-87-9
IUPAC Navn:
5-nitro-1,2-dihydroacenaphthylene
Synonymer:
5-nitroacenaphthene
Molekylvægt (g/mol):
199.21
Molekylformel:
C12H9NO2
InChi Key:
CUARLQDWYSRQDF-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23
1
–
4
af
4
Resultater
1
5-Nitroacenaphthene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Nitroacenaphthene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Nitroacenaphthene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.