missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Nitroacenaphthene, SPEX CertiPrep™

Product Code. 16389357 Shop All SPEX Certiprep Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16389357 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16389357 missing translation for 'mfr' SPEX Certiprep missing translation for 'supplierNo' S2685

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
5-Nitroacenaphthene602-87-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Kemisk navn eller materiale 5-Nitroacenaphthene
Emballage 2mL Amber Prescored Glass Ampule
CAS 602-87-9, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Metoder For US EPA Method 8270
Molekylær formel C12H9NO2
MDL nummer MFCD00014277
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle CUARLQDWYSRQDF-UHFFFAOYSA-N
SMIL [O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23
IUPAC navn 5-nitro-1,2-dihydroacenaphthylene
Opløsningsmiddel eller Matrix Methanol-P&T
Molekylvægt (g/mol) 199.21
PubChem CID 11769
Mængde 1 mL
ChEBI CHEBI:82350
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.