CAS RN 59837-11-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Acetamido-2-deoxy-4,6-O-(4-methoxybenzylidene)-3-O-(2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranosyl)-4-nitrophenyl-Alpha-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Acetamido-2-deoxy-4,6-O-(4-methoxybenzylidene)-3-O-(2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranosyl)-4-nitrophenyl-Alpha-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.