CAS RN 59262-85-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
erythro-5-Amino-4-chloro-6-(2-hydroxy-3-nonylamino)pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
erythro-5-Amino-4-chloro-6-(2-hydroxy-3-nonylamino)pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
erythro-5-Amino-4-chloro-6-(2-hydroxy-3-nonylamino)pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.