missing translation for 'onlineSavingsMsg'
Learn More

erythro-5-Amino-4-chloro-6-(2-hydroxy-3-nonylamino)pyrimidine, TRC

Product Code. 30323055
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30323055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323055 Supplier Toronto Research Chemicals Supplier No. TRCA604175100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale erythro-5-Amino-4-chloro-6-(2-hydroxy-3-nonylamino)pyrimidine
CAS 59262-85-0
Molekylær formel C13H23ClN4O
Mængde 100 mg
Synonym NSC 272455, erythro-3-(5-Amino-6-chloropyrimidin-4-ylamino)nonan-2-ol
InChI formel InChI=1S/C13H23ClN4O/c1-3-4-5-6-7-10(9(2)19)18-13-11(15)12(14)16-8-17-13/h8-10,19H,3-7,15H2,1-2H3,(H,16,17,18)/t9-,10+/m1/s1
SMIL O[C@H](C)[C@H](CCCCCC)NC1=C(N)C(Cl)=NC=N1
IUPAC navn (2R,3S)-3-((5-amino-6-chloropyrimidin-4-yl)amino)nonan-2-ol
Molekylvægt (g/mol) 286.8
Formel vægt 286.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.