CAS RN 5906-35-4
IUPAC Navn:
azepan-1-amine
Synonymer:
azepan-1-amine
Molekylvægt (g/mol):
114.19
Molekylformel:
C6H14N2
InChi Key:
UGBKOURNNQREPE-UHFFFAOYSA-N
SMILES:
NN1CCCCCC1
1
–
3
af
3
Resultater
1
N-Aminohexamethylenimine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Aminohexamethylenimine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Aminohexamethylenimine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.