CAS RN 58794-09-5
IUPAC Navn:
7-bromoisoquinoline
Synonymer:
7-bromoisoquinoline
Molekylvægt (g/mol):
208.06
Molekylformel:
C9H6BrN
InChi Key:
KABRXLINDSPGDF-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2C=CN=CC2=C1
1
–
3
af
3
Resultater
1
7-Bromoisoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Bromoisoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Bromoisoquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.