CAS RN 586-91-4
IUPAC Navn:
4-[2-(4-carboxyphenyl)diazen-1-yl]benzoic acid
Synonymer:
4-[2-(4-carboxyphenyl)diazen-1-yl]benzoic acid
Molekylvægt (g/mol):
270.24
Molekylformel:
C14H10N2O4
InChi Key:
NWHZQELJCLSKNV-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(O)=O
1
–
3
af
3
Resultater
1
Azobenzene-4,4'-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Azobenzene-4,4'-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Azobenzene-4,4'-dicarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.