CAS RN 5858-81-1
IUPAC Navn:
disodium (4E)-4-[2-(4-methyl-2-sulfonatophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxylate
Synonymer:
pigment red 57
Molekylvægt (g/mol):
430.34
Molekylformel:
C18H12N2Na2O6S
InChi Key:
CTBASIXHDUUSCV-SDXCCTRSSA-L
SMILES:
[Na+].[Na+].CC1=CC=C(N\N=C2\C(=O)C(=CC3=CC=CC=C23)C([O-])=O)C(=C1)S([O-])(=O)=O
1
–
3
af
3
Resultater
1
Disodium 3-Hydroxy-4-[(4-methyl-2-sulfonatophenyl)diazenyl]-2-naphthoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Disodium 3-Hydroxy-4-[(4-methyl-2-sulfonatophenyl)diazenyl]-2-naphthoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Disodium 3-Hydroxy-4-[(4-methyl-2-sulfonatophenyl)diazenyl]-2-naphthoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.