CAS RN 58383-83-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(5R,11S,13S)-rel-(+)-5,6,7,8,9,10,11,12-Octahydro-3-methoxy-5-methyl-5,11-methanobenzocyclodecen-13-amine L-Tartaric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(5R,11S,13S)-rel-(+)-5,6,7,8,9,10,11,12-Octahydro-3-methoxy-5-methyl-5,11-methanobenzocyclodecen-13-amine L-Tartaric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(5R,11S,13S)-rel-(+)-5,6,7,8,9,10,11,12-Octahydro-3-methoxy-5-methyl-5,11-methanobenzocyclodecen-13-amine L-Tartaric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.