CAS RN 583-57-3
IUPAC Navn:
1,2-dimethylcyclohexane
Synonymer:
1,2-dimethylcyclohexane 1,2-dimethylcyclohexane (cis)
Molekylvægt (g/mol):
112.22
Molekylformel:
C8H16
InChi Key:
KVZJLSYJROEPSQ-UHFFFAOYNA-N
SMILES:
CC1CCCCC1C
1
–
3
af
3
Resultater
1
1,2-Dimethylcyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Dimethylcyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2-Dimethylcyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.