CAS RN 582315-72-8
IUPAC Navn:
4-butoxy-5-(2,6-difluoro-4-methylbenzoyl)-1H-pyrazolo[3,4-b]pyridine
Synonymer:
4-butoxy-5-(2,6-difluoro-4-methylbenzoyl)-1H-pyrazolo[3,4-b]pyridine
Molekylvægt (g/mol):
345.35
Molekylformel:
C18H17F2N3O2
InChi Key:
SCFMWQIQBVZOQR-UHFFFAOYSA-N
SMILES:
CCCCOC1=C2C=NNC2=NC=C1C(=O)C1=C(F)C=C(C)C=C1F
1
–
2
af
2
Resultater
1
Tocris Bioscience™ BMS 265246, >97%
CAS: 582315-72-8 Molekylær formel: C18H17F2N3O2
2
BMS-265246, MedChemExpress
MedChemExpress BMS-265246 is a potent and selective cyclin-dependent kinase CDK1 and CDK2 inhibitor, with IC50 values of 6 and 9 nM, respectively. BMS-265246 inhibits CHI3L1 (chitinase 3-like-1) stimulation of ACE2 (angiotensin converting enzyme 2) and SPP (viral spike protein priming proteases). BMS-265246 can be used for ovarian cancer and COVID-19 research.