CAS RN 5809-23-4
IUPAC Navn:
2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid
Synonymer:
2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid
Molekylvægt (g/mol):
313.35
Molekylformel:
C18H19NO4
InChi Key:
FQNKTJPBXAZUGC-UHFFFAOYSA-N
SMILES:
CCN(CC)C1=CC=C(C(=O)C2=CC=CC=C2C(O)=O)C(O)=C1
1
–
3
af
3
Resultater
1
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.