CAS RN 57690-62-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester 3,4-Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester 3,4-Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester 3,4-Diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.