CAS RN 5714-73-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Methenamine Hippurate, TRC
CAS: 5714-73-8 Molekylær formel: C9 H9 N O3 . C6 H12 N4 Molekylvægt (g/mol): 319.36 Synonym: Glycine, N-benzoyl-, compd. with 1,3,5,7-tetraazatricyclo[3.3.1.13,7]decane (1:1),Hippuric acid, compd. with hexamethylenetetramine (1:1) (8CI),1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, compd. with N-benzoylglycine (1:1),Hexamethylenetetramine, monohippurate (8CI),Haiprex,Hexamethylenetetramine hippurate,Hexamethylenetetramine hippuric acid complex,Hexamine hippurate,Hippramine,Hiprex,Methenamine hippurate,R-657,Urex,Viapta SMIL: OC(=O)CNC(=O)c1ccccc1.C2N3CN4CN(CN2C4)C3