CAS RN 571-92-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
6-Ketoestradiol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6-Keto 17Beta-Estradiol, TRC
CAS: 571-92-6 Molekylær formel: C18 H22 O3 Molekylvægt (g/mol): 286.37 Synonym: 6-Ketoestradiol IUPAC navn: (8R,9S,13S,14S,17S)-3,17-dihydroxy-13-methyl-8,9,11,12,14,15,16,17-octahydro-7H-cyclopenta[a]phenanthren-6-one SMIL: C[C@]12CC[C@H]3[C@@H](CC(=O)c4cc(O)ccc34)[C@@H]1CC[C@@H]2O