CAS RN 56746-19-1
IUPAC Navn:
4-bromo-2,6-diethylaniline
Synonymer:
4-bromo-2,6-diethylaniline
Molekylvægt (g/mol):
228.13
Molekylformel:
C10H14BrN
InChi Key:
BEJYDMQQZUACPW-UHFFFAOYSA-N
SMILES:
CCC1=CC(Br)=CC(CC)=C1N
1
–
1
af
1
Resultater
1
4-Bromo-2,6-diethylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.