CAS RN 56586-13-1
IUPAC Navn:
2-methylcyclohexane-1-carboxylic acid
Synonymer:
2-methylcyclohexane-1-carboxylic acid
Molekylvægt (g/mol):
142.2
Molekylformel:
C8H14O2
InChi Key:
VSKXJRZPVDLHFY-UHFFFAOYNA-N
SMILES:
CC1CCCCC1C(O)=O
1
–
3
af
3
Resultater
1
2-Methyl-1-cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Methyl-1-cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Methyl-1-cyclohexanecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.