CAS RN 560-34-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(11Beta,16Beta)-9-Fluoro-11,17,21-trihydroxy-16-methylpregn-4-ene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11b,16b)-9-Fluoro-11,17,21-trihydroxy-16-methylpregn-4-ene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.