CAS RN 55621-49-3
IUPAC Navn:
5,6-diamino-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonymer:
5,6-diamino-1,3-dihydro-1,3-benzodiazol-2-one
Molekylvægt (g/mol):
164.17
Molekylformel:
C7H8N4O
InChi Key:
BRISYWKBJNSDPL-UHFFFAOYSA-N
SMILES:
NC1=C(N)C=C2NC(=O)NC2=C1
1
–
3
af
3
Resultater
1
5,6-Diamino-2-hydroxybenzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,6-Diamino-2-hydroxybenzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,6-Diamino-2-hydroxybenzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.