CAS RN 55290-64-7
IUPAC Navn:
5,6-dimethyl-2,3-dihydro-1λ⁶,4λ⁶-dithiine-1,1,4,4-tetrone
Synonymer:
dimethipin
Molekylvægt (g/mol):
210.26
Molekylformel:
C6H10O4S2
InChi Key:
PHVNLLCAQHGNKU-UHFFFAOYSA-N
SMILES:
CC1=C(C)S(=O)(=O)CCS1(=O)=O
1
–
1
af
1
Resultater
1
Dimethipin 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality