CAS RN 55272-86-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Cyano-N’,N' '-bis[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Cyano-N’,N' '-bis[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Cyano-N’,N' '-bis[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]guanidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.