CAS RN 55119-09-0
IUPAC Navn:
1,3,6,8-tetrabromo-9H-carbazole
Synonymer:
1,3,6,8-tetrabromo-9H-carbazole
Molekylvægt (g/mol):
482.8
Molekylformel:
C12H5Br4N
InChi Key:
FNHISDQCWYSMTO-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(NC3=C2C=C(Br)C=C3Br)C(Br)=C1
1
–
2
af
2
Resultater
1
1,3,6,8-Tetrabromocarbazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,3,6,8-Tetrabromocarbazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.