CAS RN 549-32-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Reynoutrin, MedChemExpress
MedChemExpress Reynoutrin (Quercetin-3-D-xyloside) is a flavonoid from Psidium cattleianum, with antioxidant and radical-scavenging activity.