CAS RN 548-77-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tectorigenin, MedChemExpress
MedChemExpress Tectorigenin is a plant isoflavonoid originally isolated from the dried flower of Pueraria thomsonii Benth.