CAS RN 54761-87-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-(Descyclohexylcarbonyl)-2-benzoyl Praziquantel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2-(Descyclohexylcarbonyl)-2-benzoyl Praziquantel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.