CAS RN 5464-70-0
IUPAC Navn:
ethyl 2,3-dibromo-3-phenylpropanoate
Synonymer:
ethyl 2,3-dibromo-3-phenylpropanoate
Molekylvægt (g/mol):
336.02
Molekylformel:
C11H12Br2O2
InChi Key:
CCYOCUPSKJUNMD-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(Br)C(Br)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
Ethyl 2,3-dibromo-3-phenylpropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.