CAS RN 54593-83-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Chlorethoxyfos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Chlorethoxyfos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Chlorethoxyfos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.