CAS RN 544478-19-5
IUPAC Navn:
3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonymer:
3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-4H-pyridine-3,5-dicarboxylate
Molekylvægt (g/mol):
398.42
Molekylformel:
C21H22N2O6
InChi Key:
BITHABUTZRAUGT-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1=C(C)N(CC#C)C(C)=C(C1C1=CC(=CC=C1)[N+]([O-])=O)C(=O)OC
1
–
1
af
1
Resultater
1
MRS1845, MedChemExpress
MedChemExpress MRS1845 is a selective store-operated calcium (SOC) channel inhibitor with an IC50 of 1.7 μM. MRS1845 is an ORAI1 inhibitor.