CAS RN 5436-43-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
PBDE 47 50 μg/mL in Nonane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
2,4,2',4'-Tetrabromodiphenyl Ether, TRC
CAS: 5436-43-1 Molekylær formel: C12 H6 Br4 O Molekylvægt (g/mol): 485.79 Synonym: Benzene, 2,4-dibromo-1-(2,4-dibromophenoxy)-,Benzene, 1,1'-oxybis[2,4-dibromo- (9CI),2,4-Dibromo-1-(2,4-dibromophenoxy)benzene,2,2',4,4'-Tetrabromodiphenyl ether,2,2',4,4'-Tetrabromodiphenyl oxide,2,4,2',4'-Tetrabromodiphenyl ether,BDE 47,NSC 21724,PBDE 47 IUPAC navn: 2,4-dibromo-1-(2,4-dibromophenoxy)benzene SMIL: Brc1ccc(Oc2ccc(Br)cc2Br)c(Br)c1