CAS RN 54278-05-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
5-(Ethoxycarbonyl)-2-formyl-4-methyl-1H-pyrrole-3-propanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(Ethoxycarbonyl)-2-formyl-4-methyl-1H-pyrrole-3-propanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-(Ethoxycarbonyl)-2-formyl-4-methyl-1H-pyrrole-3-propanoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.