CAS RN 54052-67-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
6-Amino-1-isobutyl-3-methyl-5-nitroso-2,4-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Amino-1-isobutyl-3-methyl-5-nitroso-2,4-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Amino-1-isobutyl-3-methyl-5-nitroso-2,4-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.