CAS RN 536-80-1
IUPAC Navn:
iodosylbenzene
Synonymer:
iodosobenzene
Molekylvægt (g/mol):
220.01
Molekylformel:
C6H5IO
InChi Key:
JYJVVHFRSFVEJM-UHFFFAOYSA-N
SMILES:
O=[I]C1=CC=CC=C1
1
–
2
af
2
Resultater
1
Iodosobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Iodosobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.