CAS RN 53448-53-6
IUPAC Navn:
(2R,3R,4S)-2-(hydroxymethyl)oxolane-3,4-diol
Synonymer:
anhydroxylitol
Molekylvægt (g/mol):
134.13
Molekylformel:
C5H10O4
InChi Key:
KZVAAIRBJJYZOW-VPENINKCSA-N
SMILES:
OC[C@H]1OC[C@H](O)[C@H]1O
1
–
2
af
2
Resultater
1
1,4-Anhydro-D-xylitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,4-Anhydro-D-xylitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.