CAS RN 532959-63-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
PF-4878691, MedChemExpress
MedChemExpress PF-4878691 (3M-852A) is a potent, orally active, and selective Toll-like receptor 7 (TLR7) agonist modelled to dissociate its antiviral and inflammatory activities.