CAS RN 5299-60-5
IUPAC Navn:
ethyl 6-hydroxyhexanoate
Synonymer:
ethyl 6-hydroxyhexanoate
Molekylvægt (g/mol):
160.21
Molekylformel:
C8H16O3
InChi Key:
HYXRUZUPCFVWAH-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCO
1
–
3
af
3
Resultater
1
Ethyl 6-Hydroxyhexanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethyl 6-Hydroxyhexanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 6-Hydroxyhexanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.