CAS RN 526-75-0
IUPAC Navn:
2,3-dimethylphenol
Synonymer:
2,3-dimethylphenol
Molekylvægt (g/mol):
122.17
Molekylformel:
C8H10O
InChi Key:
QWBBPBRQALCEIZ-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(O)=C1C
1
–
3
af
3
Resultater
1
2,3-Dimethylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3-Dimethylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3-Dimethylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.