CAS RN 52490-15-0
IUPAC Navn:
4-(2-butyl-1-benzofuran-3-carbonyl)phenol
Synonymer:
4-(2-butyl-1-benzofuran-3-carbonyl)phenol
Molekylvægt (g/mol):
294.35
Molekylformel:
C19H18O3
InChi Key:
ZHGKQUXXASLVQQ-UHFFFAOYSA-N
SMILES:
CCCCC1=C(C(=O)C2=CC=C(O)C=C2)C2=CC=CC=C2O1
1
–
4
af
4
Resultater
1
2-Butyl-3-(4-hydroxybenzoyl)benzofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Butyl-3-(4-hydroxybenzoyl)benzofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Butyl-3-(4-hydroxybenzoyl)benzofuran, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Amiodarone EP Impurity E, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.