Få mere at vide
Amiodarone EP Impurity E, Mikromol™
Artikelnummer. 30051322
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051322 25 mg
30050807 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051322 Leverandør Mikromol Leverandørnr. MM0697.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Amiodarone EP Impurity E
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Butylbenzofuran-3-yl)(4-hydroxyphenyl)methanone
CAS 52490-15-0
Analyt- eller komponentnavne (2-Butylbenzofuran-3-yl)(4-hydroxyphenyl)methanone
Forsendelsestilstand Room Temperature
Molekylær formel C19 H18 O3
Certificeringer/compliance ISO 17025
Synonym Amiodarone Hydrochloride Imp. E (EP), Amiodarone Imp. E (EP), Amiodarone USP Related Compound E, Amiodarone USP RC E, (2-Butylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, Amiodarone Related Compound E, Amiodarone Hydrochloride Impurity E, Amiodarone Impurity E
InChI formel InChI=1S/C19H18O3/c1-2-3-7-17-18(15-6-4-5-8-16(15)22-17)19(21)13-9-11-14(20)12-10-13/h4-6,8-12,20H,2-3,7H2,1H3
SMIL CCCCc1oc2ccccc2c1C(=O)c3ccc(O)cc3
IUPAC navn (2-butyl-1-benzofuran-3-yl)-(4-hydroxyphenyl)methanone
Molekylvægt (g/mol) 294.34
Mængde 25 mg
Formel vægt 294.1256 g/mol