CAS RN 52034-92-1
CAS RN 52034-92-1
IUPAC Navn:
2,2-dicyclohexylacetic acid
Synonymer:
dicyclohexylacetic acid
Molekylvægt (g/mol):
224.34
Molekylformel:
C14H24O2
InChi Key:
PGGMEZOUAPIYOY-UHFFFAOYSA-N
SMILES:
OC(=O)C(C1CCCCC1)C1CCCCC1
1
–
1
af
1
Resultater
1
Dicyclohexylacetic acid, 98+%
CAS: 52034-92-1 Molekylær formel: C14H24O2 Molekylvægt (g/mol): 224.344 MDL nummer: MFCD00075017 InChI nøgle: PGGMEZOUAPIYOY-UHFFFAOYSA-N Synonym: dicyclohexylacetic acid,hexahydro-diphenylessig,dicyclohexyl acetic acid,dicyclohexyl-acetic acid,acmc-20ala4 PubChem CID: 3661562 IUPAC navn: 2,2-dicyclohexyleddikesyre SMIL: C1CCC(CC1)C(C2CCCCC2)C(=O)O