CAS RN 51721-68-7
IUPAC Navn:
4-methoxybenzene-1-carboximidamide hydrochloride
Synonymer:
4-methoxybenzenecarboximidamide hydrochloride
Molekylvægt (g/mol):
186.64
Molekylformel:
C8H11ClN2O
InChi Key:
AJOSDIDPIBJFAI-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=C(C=C1)C(N)=N
1
–
2
af
2
Resultater
1
4-Methoxybenzamidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methoxybenzamidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.