CAS RN 51411-04-2
IUPAC Navn:
2-{2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaen-3-yl}acetic acid
Synonymer:
alrestatin 阿司他丁 alrestatinum alrestatino alrestatine
Molekylvægt (g/mol):
255.23
Molekylformel:
C14H9NO4
InChi Key:
GCUCIFQCGJIRNT-UHFFFAOYSA-N
SMILES:
OC(=O)CN1C(=O)C2=CC=CC3=CC=CC(C1=O)=C23
1
–
1
af
1
Resultater
1
Alrestatin, MedChemExpress
MedChemExpress Alrestatin is an inhibitor of aldose reductase, an enzyme involved in the pathogenesis of complications of diabetes mellitus, including diabetic neuropathy.