CAS RN 51218-49-6
IUPAC Navn:
2-chloro-N-(2,6-diethylphenyl)-N-(2-propoxyethyl)acetamide
Synonymer:
solnet pretilchlor pretilachlore pretilachlor rifit
Molekylvægt (g/mol):
311.85
Molekylformel:
C17H26ClNO2
InChi Key:
YLPGTOIOYRQOHV-UHFFFAOYSA-N
SMILES:
CCCOCCN(C(=O)CCl)C1=C(CC)C=CC=C1CC
1
–
3
af
3
Resultater
1
Pretilachlor, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Pretilachlor, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pretilachlor, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.