CAS RN 51077-01-1
IUPAC Navn:
1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylic acid
Synonymer:
tosyl-D-proline
Molekylvægt (g/mol):
269.32
Molekylformel:
C12H15NO4S
InChi Key:
CGPHGPCHVUSFFA-UHFFFAOYNA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)N1CCCC1C(O)=O
1
–
3
af
3
Resultater
1
1-[(4-Methylphenyl)sulfonyl]proline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[(4-Methylphenyl)sulfonyl]proline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-[(4-Methylphenyl)sulfonyl]proline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.