CAS RN 509093-47-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Tocris Bioscience™ IRAK1/4 Inhibitor I
IRAK4 and IRAK1 inhibitor
2
IRAK-1-4 Inhibitor I, MedChemExpress
MedChemExpress IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor-associated kinase 1/4 (IRAK 1/4) with IC50s of 0.2 μM and 0.3 μM, respectively.
3
4